siRNA Details
| Gene Symbol: Aig1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134425.1 | NP_001127897.1 | 5 | 675 | ||
| XM_006227642.2 | XP_006227704.1 | 5 | 874 | |||
| GenBank mRNA | BC167098.1 | - | 5 | 675 | 7319120 | |
| FQ229550.1 | - | 5 | 693 | |||
| FQ229983.1 | - | 5 | 691 | |||

Aig1
Genomic Map