siRNA Details
| Gene Symbol: LOC103691059 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008759084.1 | XP_008757306.1 | 1 | 816 | ||
| Gene Symbol: Zfp418 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191620.1 | NP_001178549.1 | 4 | 1596 | ||
| XM_006228177.2 | XP_006228239.1 | 3 | 1740 | |||
| GenBank mRNA | FQ212491.1 | - | 4 | 1690 | ||

LOC103691059
Genomic Map