siRNA Details
| Gene Symbol: Zscan5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_218239.5 | XP_218239.5 | 4 | 960 | ||
| Gene Symbol: LOC100912263 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008759488.1 | XP_008757710.1 | 1 | 236 | ||

Zscan5b
Genomic Map