siRNA Details
| Gene Symbol: Vom1r33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008942.1 | NP_001008942.1 | 1 | 375 | ||
| GenBank mRNA | AY510340.1 | - | 1 | 375 | ||
| Gene Symbol: Vom1r39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001167551.1 | NP_001161023.1 | 1 | 375 | ||

Vom1r33
Genomic Map