siRNA Details
| Gene Symbol: Ube2s | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106224.2 | NP_001099694.1 | 3 | 381 | ||
| GenBank mRNA | BC169077.1 | - | 3 | 381 | 5620985 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ube2s| Gene Symbol: Ube2s | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106224.2 | NP_001099694.1 | 3 | 381 | ||
| GenBank mRNA | BC169077.1 | - | 3 | 381 | 5620985 | |