siRNA Details
| Gene Symbol: Lilrb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013894.1 | NP_001013916.1 | 4 | 488 | ||
| XM_006228238.2 | XP_006228300.1 | 4 | 559 | |||
| XM_006228239.2 | XP_006228301.1 | 4 | 559 | |||
| GenBank mRNA | AY359280.1 | - | 4 | 488 | ||
| AY359281.1 | - | 4 | 488 | |||

Lilrb4
Genomic Map