siRNA Details
| Gene Symbol: Lilrb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013894.1 | NP_001013916.1 | 10 | 970 | ||
| XM_006228238.2 | XP_006228300.1 | 11 | 1158 | |||
| GenBank mRNA | AY359280.1 | - | 11 | 1087 | ||
| AY359281.1 | - | 10 | 970 | |||

Lilrb4
Genomic Map