siRNA Details
| Gene Symbol: Gltscr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207591.1 | NP_997474.1 | 2, 3 | 297 | ||
| GenBank mRNA | AY535002.1 | - | 2, 3 | 297 | ||
| BC086359.1 | - | 2, 3 | 301 | 7304013 | ||
| FQ212888.1 | - | 2, 3 | 328 | |||
| FQ213201.1 | - | 2, 3 | 307 | |||
| FQ218937.1 | - | 2, 3 | 303 | |||

Gltscr2
Genomic Map