siRNA Details
| Gene Symbol: LOC102553890 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006228355.2 | XP_006228417.2 | 3 | 318 | ||
| Gene Symbol: LOC102551565 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006228341.2 | XP_006228403.1 | 2 | 691 | ||

LOC102553890
Genomic Map