siRNA Details
Gene Symbol: LOC102551565 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006228341.2 | XP_006228403.1 | 1 | 522 |
Gene Symbol: LOC102553890 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006228355.2 | XP_006228417.2 | 2 | 149 |
