siRNA Details
| Gene Symbol: Prkd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013895.1 | NP_001013917.1 | 16 | 2374 | ||
| XM_008758905.1 | XP_008757127.1 | 16 | 3261 | |||
| GenBank mRNA | BC083592.1 | - | 16 | 2374 | 7132731 | |
| FQ218070.1 | - | 17 | 2465 | |||

Prkd2
Genomic Map