siRNA Details
| Gene Symbol: Psg29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025641.1 | NP_001020812.1 | 3 | 718 | ||
| GenBank mRNA | BC097354.1 | - | 3 | 718 | 7441714 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Psg29| Gene Symbol: Psg29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025641.1 | NP_001020812.1 | 3 | 718 | ||
| GenBank mRNA | BC097354.1 | - | 3 | 718 | 7441714 | |