siRNA Details
| Gene Symbol: Mill1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011931.2 | NP_001011931.1 | 5 | 1052 | ||
| XM_008758906.1 | XP_008757128.1 | 6 | 1115 | |||
| XM_008758907.1 | XP_008757129.1 | 5 | 1138 | |||
| GenBank mRNA | AB113960.1 | - | 5 | 1061 | ||
| AB113962.1 | - | 5 | 1052 | |||

Mill1
Genomic Map