siRNA Details
| Gene Symbol: Ercc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106228.1 | NP_001099698.1 | 7 | 745 | ||
| XM_006228395.2 | XP_006228457.1 | 7 | 763 | |||
| XM_006228396.1 | XP_006228458.1 | 7 | 723 | |||
| GenBank mRNA | FQ225470.1 | - | 7 | 734 | ||

Ercc1
Genomic Map