siRNA Details
| Gene Symbol: Clptm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106232.1 | NP_001099702.1 | 8 | 1000 | ||
| GenBank mRNA | BC166760.1 | - | 8 | 993 | 7930858 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Clptm1| Gene Symbol: Clptm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106232.1 | NP_001099702.1 | 8 | 1000 | ||
| GenBank mRNA | BC166760.1 | - | 8 | 993 | 7930858 | |