siRNA Details
| Gene Symbol: Apoc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085352.1 | NP_001078821.1 | 3, 4 | 244 | ||
| GenBank mRNA | FQ209611.1 | - | 3, 4 | 323 | ||
| FQ210391.1 | - | 3, 4 | 253 | |||
| FQ210737.1 | - | 3, 4 | 254 | |||
| FQ211227.1 | - | 3, 4 | 254 | |||
| FQ212251.1 | - | 3, 4 | 254 | |||
| FQ218101.1 | - | 3, 4 | 254 | |||
| FQ218174.1 | - | 3, 4 | 254 | |||
| FQ218270.1 | - | 3, 4 | 253 | |||

Apoc2
Genomic Map