siRNA Details
| Gene Symbol: Tescl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106233.1 | NP_001099703.1 | 1 | 153 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tescl| Gene Symbol: Tescl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106233.1 | NP_001099703.1 | 1 | 153 | ||