siRNA Details
| Gene Symbol: Irgq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135742.2 | NP_001129214.2 | 3 | 2235 | ||
| XM_006228410.2 | XP_006228472.1 | 2 | 2193 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Irgq| Gene Symbol: Irgq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135742.2 | NP_001129214.2 | 3 | 2235 | ||
| XM_006228410.2 | XP_006228472.1 | 2 | 2193 | |||