siRNA Details
Gene Symbol: LOC100909620 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003748782.3 | XP_003748830.1 | 4 | 585 | ||
XM_006228510.2 | XP_006228572.1 | 4 | 582 | |||
XM_006228511.2 | XP_006228573.1 | 4 | 582 | |||
XM_008759059.1 | XP_008757281.1 | 4 | 590 | |||
XM_008759060.1 | XP_008757282.1 | 4 | 443 |
Gene Symbol: LOC100910565 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006228509.2 | XP_006228571.1 | 309 |
Gene Symbol: RGD1564380 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170329.1 | NP_001163800.1 | 257 | |||
XM_006228544.2 | XP_006228606.1 | 4 | 582 |
