siRNA Details
| Gene Symbol: Ccdc97 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106235.1 | NP_001099705.1 | 2, 3 | 660 | ||
| GenBank mRNA | FQ212311.1 | - | 2, 3 | 584 | ||
| FQ232330.1 | - | 2, 3 | 580 | |||
| FQ235099.1 | - | 2, 3 | 580 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc97| Gene Symbol: Ccdc97 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106235.1 | NP_001099705.1 | 2, 3 | 660 | ||
| GenBank mRNA | FQ212311.1 | - | 2, 3 | 584 | ||
| FQ232330.1 | - | 2, 3 | 580 | |||
| FQ235099.1 | - | 2, 3 | 580 | |||