siRNA Details
| Gene Symbol: Blvrb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106236.1 | NP_001099706.1 | 2, 3 | 388 | ||
| XM_008759117.1 | XP_008757339.1 | 2, 3 | 309 | |||
| GenBank mRNA | BC168943.1 | - | 2, 3 | 292 | 7322093 | |
| FQ228106.1 | - | 2, 3 | 272 | |||
| FQ232598.1 | - | 2, 3 | 335 | |||

Blvrb
Genomic Map