siRNA Details
| Gene Symbol: Map4k1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106243.1 | NP_001099713.1 | 28 | 2267 | ||
| XM_006228716.2 | XP_006228778.1 | 28 | 3226 | |||
| XM_008759122.1 | XP_008757344.1 | 26 | 3091 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Map4k1| Gene Symbol: Map4k1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106243.1 | NP_001099713.1 | 28 | 2267 | ||
| XM_006228716.2 | XP_006228778.1 | 28 | 3226 | |||
| XM_008759122.1 | XP_008757344.1 | 26 | 3091 | |||