siRNA Details
| Gene Symbol: Ggn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013065.1 | NP_001013083.1 | 1 | 1404 | ||
| XM_006228718.2 | XP_006228780.1 | 2 | 1999 | |||
| XM_006228719.2 | XP_006228781.1 | 2 | 1833 | |||
| XM_008759123.1 | XP_008757345.1 | 2 | 2159 | |||
| GenBank mRNA | BC081922.1 | - | 1 | 1404 | 7112701 | |
| BC100147.1 | - | 2 | 1148 | 7111711 | ||

Ggn
Genomic Map