siRNA Details
| Gene Symbol: Spint2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001082549.1 | NP_001076018.1 | 7 | 937 | ||
| NM_199087.1 | NP_954518.1 | 6 | 766 | |||
| GenBank mRNA | BC061768.1 | - | 6 | 766 | 5621353 | |
| FQ229514.1 | - | 7 | 665 | |||

Spint2
Genomic Map