siRNA Details
| Gene Symbol: Lin37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106245.1 | NP_001099715.1 | 5, 6 | 448 | ||
| XM_006228769.2 | XP_006228831.1 | 5, 6 | 869 | |||
| GenBank mRNA | FQ233946.1 | - | 5, 6 | 416 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lin37| Gene Symbol: Lin37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106245.1 | NP_001099715.1 | 5, 6 | 448 | ||
| XM_006228769.2 | XP_006228831.1 | 5, 6 | 869 | |||
| GenBank mRNA | FQ233946.1 | - | 5, 6 | 416 | ||