siRNA Details
| Gene Symbol: Tmem147 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001038494.1 | NP_001033583.1 | 6 | 648 | ||
| GenBank mRNA | BC111082.1 | - | 6 | 648 | 7455198 | |
| FQ234356.1 | - | 6 | 663 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem147| Gene Symbol: Tmem147 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001038494.1 | NP_001033583.1 | 6 | 648 | ||
| GenBank mRNA | BC111082.1 | - | 6 | 648 | 7455198 | |
| FQ234356.1 | - | 6 | 663 | |||