siRNA Details
Gene Symbol: Pepd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009641.1 | NP_001009641.1 | 2 | 183 | ||
GenBank mRNA | BC088440.1 | - | 2 | 183 | 7126171 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Pepd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009641.1 | NP_001009641.1 | 2 | 183 | ||
GenBank mRNA | BC088440.1 | - | 2 | 183 | 7126171 |