siRNA Details
| Gene Symbol: Pepd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009641.1 | NP_001009641.1 | 12 | 961 | ||
| GenBank mRNA | BC088440.1 | - | 12 | 961 | 7126171 | |
| FQ235015.1 | - | 1 | 23 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pepd| Gene Symbol: Pepd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009641.1 | NP_001009641.1 | 12 | 961 | ||
| GenBank mRNA | BC088440.1 | - | 12 | 961 | 7126171 | |
| FQ235015.1 | - | 1 | 23 | |||