siRNA Details
| Gene Symbol: Cep89 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127599.1 | NP_001121071.1 | 14 | 1594 | ||
| XM_006228881.2 | XP_006228943.1 | 14 | 1663 | |||
| XM_006228883.2 | XP_006228945.1 | 14 | 1663 | |||
| GenBank mRNA | BC161945.1 | - | 14 | 1594 | 9025363 | |

Cep89
Genomic Map