siRNA Details
| Gene Symbol: LOC100912350 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003748841.3 | XP_003748889.1 | 1372 | |||
| XM_006228898.2 | XP_006228960.1 | 1372 | ||||
| Gene Symbol: Zfp507 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106248.1 | NP_001099718.2 | 3 | 909 | ||
| XM_006228885.2 | XP_006228947.1 | 3 | 870 | |||
| XM_006228886.2 | XP_006228948.1 | 3 | 941 | |||

LOC100912350
Genomic Map