siRNA Details
| Gene Symbol: Zfp536 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008759267.1 | XP_008757489.1 | 4 | 2576 | ||
| XM_008759268.1 | XP_008757490.1 | 4 | 2411 | |||
| XM_008759269.1 | XP_008757491.1 | 4 | 2370 | |||
| XM_008759270.1 | XP_008757492.1 | 4 | 2172 | |||
| XM_008759271.1 | XP_008757493.1 | 4 | 2335 | |||
| XM_008759272.1 | XP_008757494.1 | 4 | 1626 | |||
| XM_008759273.1 | XP_008757495.1 | 4 | 1630 | |||
| XM_008759274.1 | XP_008757496.1 | 4 | 2211 | |||
| XM_008759275.1 | XP_008757497.1 | 4 | 1728 | |||

Zfp536
Genomic Map