siRNA Details
Gene Symbol: Klk1c12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005382.1 | NP_001005382.1 | 3 | 261 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Klk1c12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005382.1 | NP_001005382.1 | 3 | 261 |