siRNA Details
| Gene Symbol: RGD1309036 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106255.1 | NP_001099725.1 | 3 | 437 | ||
| XM_006228989.2 | XP_006229051.1 | 3 | 619 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1309036| Gene Symbol: RGD1309036 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106255.1 | NP_001099725.1 | 3 | 437 | ||
| XM_006228989.2 | XP_006229051.1 | 3 | 619 | |||