siRNA Details
| Gene Symbol: Aspdh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009643.2 | NP_001009643.2 | 3 | 301 | ||
| XM_006228995.1 | XP_006229057.1 | 3 | 301 | |||
| GenBank mRNA | BC088252.1 | - | 3 | 301 | 7372248 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Aspdh| Gene Symbol: Aspdh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009643.2 | NP_001009643.2 | 3 | 301 | ||
| XM_006228995.1 | XP_006229057.1 | 3 | 301 | |||
| GenBank mRNA | BC088252.1 | - | 3 | 301 | 7372248 | |