siRNA Details
| Gene Symbol: Josd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106256.2 | NP_001099726.2 | 3 | 331 | ||
| XM_006228996.2 | XP_006229058.1 | 4 | 613 | |||
| XM_006228997.2 | XP_006229059.1 | 4 | 610 | |||
| XM_006228998.2 | XP_006229060.1 | 3 | 552 | |||
| GenBank mRNA | DV214517.1 | - | 3 | 331 | ||

Josd2
Genomic Map