siRNA Details
| Gene Symbol: Emc10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004221.1 | NP_001004221.1 | 4 | 442 | ||
| XM_006229000.2 | XP_006229062.1 | 4 | 458 | |||
| XM_006229001.2 | XP_006229063.1 | 4 | 458 | |||
| GenBank mRNA | BC079041.1 | - | 4 | 442 | 7114835 | |

Emc10
Genomic Map