siRNA Details
| Gene Symbol: Akt1s1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106259.3 | NP_001099729.2 | 2 | 489 | ||
| XM_006229003.2 | XP_006229065.1 | 2 | 965 | |||
| XM_006229004.2 | XP_006229066.1 | 2 | 665 | |||
| XM_006229005.2 | XP_006229067.1 | 2 | 641 | |||
| GenBank mRNA | DY316571.1 | - | 2 | 685 | ||
| EV773331.1 | - | 1 | 405 | |||

Akt1s1
Genomic Map