siRNA Details
| Gene Symbol: Ptov1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008304.1 | NP_001008305.1 | 11 | 1267 | ||
| GenBank mRNA | BC085836.1 | - | 11 | 1267 | 7135127 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ptov1| Gene Symbol: Ptov1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008304.1 | NP_001008305.1 | 11 | 1267 | ||
| GenBank mRNA | BC085836.1 | - | 11 | 1267 | 7135127 | |