siRNA Details
| Gene Symbol: Tsks | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199088.1 | NP_954519.1 | 3 | 491 | ||
| XM_006229014.2 | XP_006229076.1 | 3 | 553 | |||
| XM_006229015.2 | XP_006229077.1 | 3 | 553 | |||
| XM_006229016.2 | XP_006229078.1 | 3 | 414 | |||
| XM_006229017.2 | XP_006229079.1 | 3 | 552 | |||
| XM_006229018.2 | XP_006229080.1 | 3 | 552 | |||
| XM_006229019.2 | XP_006229081.1 | 3 | 552 | |||
| XM_006229020.2 | XP_006229082.1 | 3 | 552 | |||
| GenBank mRNA | AY346104.1 | - | 3 | 491 | ||

Tsks
Genomic Map