siRNA Details
| Gene Symbol: Pih1d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024868.1 | NP_001020039.1 | 4, 5 | 726 | ||
| XM_006229027.1 | XP_006229089.1 | 5, 6 | 463 | |||
| XM_006229028.2 | XP_006229090.1 | 5, 6 | 699 | |||
| GenBank mRNA | BC097946.1 | - | 4, 5 | 726 | 7366310 | |

Pih1d1
Genomic Map