siRNA Details
| Gene Symbol: Hrc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181369.2 | NP_852034.1 | 1 | 1636 | ||
| GenBank mRNA | AY277279.1 | - | 1 | 1636 | ||
| FQ224148.1 | - | 1 | 1722 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hrc| Gene Symbol: Hrc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181369.2 | NP_852034.1 | 1 | 1636 | ||
| GenBank mRNA | AY277279.1 | - | 1 | 1636 | ||
| FQ224148.1 | - | 1 | 1722 | |||