siRNA Details
| Gene Symbol: Rasip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106261.2 | NP_001099731.1 | 10 | 2017 | ||
| XM_006229030.2 | XP_006229092.1 | 12 | 3102 | |||
| GenBank mRNA | BC168876.1 | - | 10 | 2017 | 7135603 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rasip1| Gene Symbol: Rasip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106261.2 | NP_001099731.1 | 10 | 2017 | ||
| XM_006229030.2 | XP_006229092.1 | 12 | 3102 | |||
| GenBank mRNA | BC168876.1 | - | 10 | 2017 | 7135603 | |