siRNA Details
| Gene Symbol: Sult2b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039665.1 | NP_001034754.1 | 1 | 200 | ||
| GenBank mRNA | AY827147.1 | - | 1 | 200 | ||
| BC127438.1 | - | 874 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sult2b1| Gene Symbol: Sult2b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039665.1 | NP_001034754.1 | 1 | 200 | ||
| GenBank mRNA | AY827147.1 | - | 1 | 200 | ||
| BC127438.1 | - | 874 | ||||