siRNA Details
| Gene Symbol: Htatip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106263.2 | NP_001099733.1 | 1 | 100 | ||
| XM_006229249.2 | XP_006229311.1 | 2 | 251 | |||
| XM_006229250.2 | XP_006229312.1 | 2 | 155 | |||
| XM_006229251.1 | XP_006229313.1 | 2 | 166 | |||
| GenBank mRNA | BC158803.1 | - | 1 | 100 | 7321759 | |

Htatip2
Genomic Map