siRNA Details
| Gene Symbol: Perp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106265.2 | NP_001099735.1 | 3 | 488 | ||
| GenBank mRNA | BC166608.1 | - | 3 | 488 | 5599021 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Perp| Gene Symbol: Perp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106265.2 | NP_001099735.1 | 3 | 488 | ||
| GenBank mRNA | BC166608.1 | - | 3 | 488 | 5599021 | |