siRNA Details
| Gene Symbol: UPK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006760.3 | NP_006751.1 | 5 | 854 | ||
| GenBank mRNA | AF000562.1 | - | 1 | 367 | ||
| Y13645.1 | - | 5 | 839 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

UPK2| Gene Symbol: UPK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006760.3 | NP_006751.1 | 5 | 854 | ||
| GenBank mRNA | AF000562.1 | - | 1 | 367 | ||
| Y13645.1 | - | 5 | 839 | |||