siRNA Details
| Gene Symbol: UPK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006760.3 | NP_006751.1 | 3 | 309 | ||
| GenBank mRNA | AB009454.1 | - | 3 | 196 | ||
| AY455312.1 | - | 3 | 256 | |||
| BC113900.1 | - | 3 | 287 | 40029623 | ||
| CD556028.1 | - | 3 | 324 | |||
| Y13645.1 | - | 3 | 294 | |||

UPK2
Genomic Map