siRNA Details
| Gene Symbol: Chsy1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106268.1 | NP_001099738.1 | 3 | 1032 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Chsy1| Gene Symbol: Chsy1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106268.1 | NP_001099738.1 | 3 | 1032 | ||