siRNA Details
Gene Symbol: Mrpl46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013068.1 | NP_001013086.1 | 3 | 465 | ||
GenBank mRNA | BC086407.1 | - | 3 | 465 | 7306696 | |
FQ212933.1 | - | 3 | 461 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Mrpl46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013068.1 | NP_001013086.1 | 3 | 465 | ||
GenBank mRNA | BC086407.1 | - | 3 | 465 | 7306696 | |
FQ212933.1 | - | 3 | 461 |