siRNA Details
| Gene Symbol: Fam103a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127451.1 | NP_001120923.1 | 3 | 299 | ||
| GenBank mRNA | BC089896.1 | - | 2 | 948 | 7381229 | |
| DV717487.1 | - | 2 | 152 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam103a1| Gene Symbol: Fam103a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127451.1 | NP_001120923.1 | 3 | 299 | ||
| GenBank mRNA | BC089896.1 | - | 2 | 948 | 7381229 | |
| DV717487.1 | - | 2 | 152 | |||